siRNA Details
| Gene Symbol: Fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081253.1 | NP_001074722.1 | 4 | 2180 | ||
| GenBank mRNA | AK019714.1 | - | 4 | 2118 | ||
| AK030012.1 | - | 4 | 2001 | |||
| BC026503.1 | - | 3 | 474 | |||
| BC098484.1 | - | 4 | 2180 | 30053208 | ||

Fbxo43
Genomic Map