siRNA Details
| Gene Symbol: Stxbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081344.2 | NP_001074813.2 | 22, 23 | 3160 | ||
| GenBank mRNA | AF516607.1 | - | 18, 19 | 2239 | ||
| AK019788.1 | - | 5, 6 | 901 | |||
| AK029982.1 | - | 3, 4 | 2911 | |||
| AK122417.1 | - | 1, 2 | 239 | |||
| AK136998.1 | - | 1, 2 | 1174 | |||
| BC038042.1 | - | 2, 3 | 528 | |||
| BC094582.1 | - | 15, 16 | 1985 | |||

Stxbp5
Genomic Map