siRNA Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024282.3 | NP_077244.1 | 2 | 566 | ||
| GenBank mRNA | AK020014.1 | - | 2 | 451 | ||
| AK029574.1 | - | 3 | 640 | |||
| AK049579.1 | - | 2 | 335 | |||
| AK146492.1 | - | 2 | 299 | |||
| AK160733.1 | - | 2 | 155 | |||
| AK216786.1 | - | 1 | 38 | |||
| BC002200.1 | - | 2 | 235 | 3489351 | ||
| BC046816.1 | - | 2 | 273 | 5702476 | ||

Desi2
Genomic Map