siRNA Details
| Gene Symbol: P2ry10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172435.3 | NP_766023.1 | 3 | 1234 | ||
| GenBank mRNA | AK020001.1 | - | 3 | 1113 | ||
| AK038124.1 | - | 2 | 1161 | |||
| AK041351.1 | - | 3 | 1250 | |||
| AK088234.1 | - | 3 | 1235 | |||
| BC053707.1 | - | 3 | 1065 | 30072257 | ||

P2ry10
Genomic Map