siRNA Details
| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172096.1 | NP_001165567.1 | 4 | 949 | ||
| NM_001172097.1 | NP_001165568.1 | 4 | 949 | |||
| NM_030197.2 | NP_084473.1 | 4 | 949 | |||
| GenBank mRNA | AK019216.1 | - | 4 | 883 | ||
| AK036606.1 | - | 4 | 833 | |||
| AK037816.1 | - | 1 | 1798 | |||
| AK051314.1 | - | 1 | 1159 | |||
| AK147382.1 | - | 4 | 952 | |||
| AK169791.1 | - | 4 | 825 | |||
| AK171596.1 | - | 4 | 325 | |||
| BC013270.1 | - | 4 | 496 | |||
| BC026363.1 | - | 4 | 919 | 4223313 | ||

Cacul1
Genomic Map