siRNA Details
| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172096.1 | NP_001165567.1 | 5 | 1095 | ||
| NM_001172097.1 | NP_001165568.1 | 5 | 1095 | |||
| NM_030197.2 | NP_084473.1 | 5 | 1095 | |||
| GenBank mRNA | AA237160.1 | - | 1 | 35 | ||
| AK019216.1 | - | 5 | 1029 | |||
| AK036606.1 | - | 5 | 979 | |||
| AK147382.1 | - | 5 | 1098 | |||
| AK169791.1 | - | 5 | 971 | |||
| AK171596.1 | - | 5 | 471 | |||
| BC013270.1 | - | 5 | 642 | |||
| BC026363.1 | - | 5 | 1065 | 4223313 | ||

Cacul1
Genomic Map