siRNA Details
| Gene Symbol: Igsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207205.2 | NP_997088.2 | 7 | 2680 | ||
| GenBank mRNA | AK045721.1 | - | 4 | 1410 | ||
| AK084011.1 | - | 7 | 2718 | |||
| AK129151.1 | - | 7 | 2680 | |||
| BC052892.1 | - | 6 | 2104 | 30055632 | ||

Igsf3
Genomic Map