siRNA Details
| Gene Symbol: Gas2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190408.1 | NP_001177337.1 | 6 | 2049 | ||
| NM_144560.3 | NP_653146.1 | 7 | 2307 | |||
| GenBank mRNA | AF508323.1 | - | 4 | 2282 | ||
| AF508324.1 | - | 5 | 1951 | |||
| AK052229.1 | - | 7 | 2312 | |||
| AK214395.1 | - | 1 | 146 | |||
| BC031785.1 | - | 6 | 1999 | 4194333 | ||

Gas2l1
Genomic Map