siRNA Details
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030235.1 | NP_084511.1 | 10 | 1386 | ||
| GenBank mRNA | AK020003.1 | - | 10 | 1363 | ||
| AK083605.1 | - | 10 | 1343 | |||
| AK122228.1 | - | 10 | 1364 | |||
| AK147365.1 | - | 10 | 1388 | |||
| BC051177.1 | - | 10 | 1402 | |||
| BC117878.1 | - | 10 | 1348 | 40095961 | ||
| BC117879.1 | - | 10 | 1348 | |||

Avl9
Genomic Map