siRNA Details
| Gene Symbol: Spsb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163750.1 | NP_001157222.1 | 7 | 1409 | ||
| NM_001163751.1 | NP_001157223.1 | 7 | 1064 | |||
| NM_027141.2 | NP_081417.2 | 7 | 1085 | |||
| GenBank mRNA | AF403038.1 | - | 7 | 1085 | ||
| AK009325.1 | - | 7 | 1085 | |||
| AK076170.1 | - | 7 | 1416 | |||
| AK082768.1 | - | 7 | 1008 | |||
| AK156579.1 | - | 6 | 1422 | |||
| AK167474.1 | - | 6 | 1402 | |||
| AK220234.1 | - | 7 | 1347 | |||
| BC026523.1 | - | 7 | 1750 | |||
| BC072556.1 | - | 7 | 1370 | 5691114 | ||

Spsb3
Genomic Map