siRNA Details
| Gene Symbol: Tnfrsf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ278264.1 | - | 1, 2 | 60 | ||
| AK154786.1 | - | 1, 2 | 86 | |||
| AK170037.1 | - | 1, 2 | 89 | |||
| AY046550.1 | - | 1, 2 | 69 | |||
| AY165625.1 | - | 1, 2 | 75 | |||
| BC106828.1 | - | 1, 2 | 71 | 40044453 | ||
| BC106829.1 | - | 1, 2 | 75 | 40044455 | ||
| BG077775.2 | - | 1, 2 | 61 | |||
| BY036522.1 | - | 1, 2 | 85 | |||
| BY201915.1 | - | 1, 2 | 91 | |||

Tnfrsf23
Genomic Map