siRNA Details
| Gene Symbol: Trim39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024468.2 | NP_077788.2 | 2 | 455 | ||
| NM_178281.1 | NP_840065.1 | 2 | 440 | |||
| GenBank mRNA | AB046382.1 | - | 2 | 455 | ||
| AK053507.1 | - | 2 | 392 | |||
| AK053591.1 | - | 1 | 3528 | |||
| AK145861.1 | - | 2 | 391 | |||
| AK166148.1 | - | 1 | 29 | |||
| AK220298.1 | - | 2 | 376 | |||
| BC031540.1 | - | 1 | 497 | 3666785 | ||

Trim39
Genomic Map