siRNA Details
| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271768.1 | NP_001258697.1 | 3 | 296 | ||
| NM_024469.2 | NP_077789.1 | 3 | 296 | |||
| GenBank mRNA | AA013582.1 | - | 3 | 297 | ||
| AB044090.1 | - | 3 | 216 | |||
| BC023231.1 | - | 3 | 248 | |||
| BC037742.1 | - | 3 | 256 | |||
| BC051555.1 | - | 3 | 257 | |||

Bhlhe41
Genomic Map