siRNA Details
| Gene Symbol: BC005537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024473.3 | NP_077793.1 | 2 | 1974 | ||
| GenBank mRNA | AK035718.1 | - | 2 | 1974 | ||
| AK050530.1 | - | 769 | ||||
| AK078765.1 | - | 2 | 1640 | |||
| AK160094.1 | - | 931 | ||||
| BC002217.1 | - | 348 | 3490515 | |||
| BC005537.1 | - | 1611 | ||||

BC005537
Genomic Map