siRNA Details
| Gene Symbol: Ublcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024475.5 | NP_077795.2 | 6 | 689 | ||
| GenBank mRNA | AK043952.1 | - | 6 | 596 | ||
| AK050031.1 | - | 6 | 600 | |||
| AK147744.1 | - | 6 | 637 | |||
| AK153998.1 | - | 6 | 601 | |||
| BC083331.1 | - | 4 | 350 | |||
| BC085111.1 | - | 5 | 592 | 30735741 | ||
| BC106093.1 | - | 5 | 566 | 5098953 | ||

Ublcp1
Genomic Map