siRNA Details
| Gene Symbol: Wbscr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024479.2 | NP_077799.2 | 5, 6 | 633 | ||
| GenBank mRNA | AK042578.1 | - | 4, 5 | 431 | ||
| AK081683.1 | - | 5, 6 | 652 | |||
| AK132481.1 | - | 5, 6 | 622 | |||
| AY138964.1 | - | 5, 6 | 633 | |||
| AY354925.2 | - | 5, 6 | 633 | |||
| AY354926.1 | - | 5, 6 | 534 | |||
| AY354927.1 | - | 4, 5 | 540 | |||

Wbscr27
Genomic Map