siRNA Details
| Gene Symbol: Cttnbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163332.1 | NP_001156804.1 | 6 | 1215 | ||
| NM_001163333.1 | NP_001156805.1 | 6 | 1207 | |||
| NM_030249.4 | NP_084525.1 | 6 | 1323 | |||
| GenBank mRNA | AK029901.1 | - | 6 | 1324 | ||
| AK030438.1 | - | 6 | 1188 | |||
| AK129359.1 | - | 6 | 1213 | |||
| AK145085.1 | - | 6 | 1189 | |||
| BC003236.1 | - | 6 | 1177 | 3498078 | ||
| BC006952.1 | - | 2 | 665 | |||

Cttnbp2nl
Genomic Map