siRNA Details
| Gene Symbol: Smim12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030252.2 | NP_084528.1 | 2 | 327 | ||
| GenBank mRNA | AK075968.1 | - | 2 | 302 | ||
| AK131915.1 | - | 2 | 292 | |||
| AK141221.1 | - | 2 | 261 | |||
| BC147067.1 | - | 2 | 289 | 9056070 | ||
| BC147068.1 | - | 2 | 289 | 9056074 | ||
| BY237825.1 | - | 2 | 328 | |||

Smim12
Genomic Map