siRNA Details
| Gene Symbol: Smim12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030252.2 | NP_084528.1 | 2 | 951 | ||
| GenBank mRNA | AK131844.1 | - | 1 | 429 | ||
| AK131915.1 | - | 2 | 916 | |||
| AK141221.1 | - | 2 | 885 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim12| Gene Symbol: Smim12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030252.2 | NP_084528.1 | 2 | 951 | ||
| GenBank mRNA | AK131844.1 | - | 1 | 429 | ||
| AK131915.1 | - | 2 | 916 | |||
| AK141221.1 | - | 2 | 885 | |||