siRNA Details
| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030254.3 | NP_084530.3 | 762 | |||
| GenBank mRNA | AK028182.1 | - | 738 | |||
| AK088616.1 | - | 776 | ||||
| AK132766.1 | - | 785 | ||||
| AK168664.1 | - | 720 | ||||
| AK168961.1 | - | 720 | ||||
| BC003311.1 | - | 603 | ||||
| BC069898.1 | - | 762 | 6489807 | |||

Tusc3
Genomic Map