siRNA Details
| Gene Symbol: Kcnip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030265.3 | NP_084541.3 | 1, 2 | 185 | ||
| GenBank mRNA | AF453243.1 | - | 1, 2 | 93 | ||
| AK039048.1 | - | 1, 2 | 186 | |||
| AK049662.1 | - | 1, 2 | 137 | |||
| AK165773.1 | - | 1, 2 | 127 | |||
| DQ148503.1 | - | 1, 2 | 137 | |||

Kcnip4
Genomic Map