siRNA Details
| Gene Symbol: Tusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019742.4 | NP_062716.1 | 2 | 290 | ||
| GenBank mRNA | AF123387.1 | - | 2 | 177 | ||
| AK004430.1 | - | 2 | 250 | |||
| AK082684.1 | - | 2 | 291 | |||
| BC048477.1 | - | 2 | 290 | 6772171 | ||

Tusc2
Genomic Map