siRNA Details
| Gene Symbol: Herpud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020586.2 | NP_065611.1 | 7 | 1834 | ||
| GenBank mRNA | AB041580.1 | - | 7 | 1629 | ||
| AK019862.1 | - | 7 | 1779 | |||
| AK135935.1 | - | 7 | 1835 | |||
| AK180080.1 | - | 45 | ||||
| AK208269.1 | - | 1 | 45 | |||
| AK209268.1 | - | 1 | 45 | |||
| AK209694.1 | - | 1 | 45 | |||
| AK214130.1 | - | 1 | 45 | |||
| BC029691.1 | - | 7 | 1052 | 5375603 | ||
| BC043693.1 | - | 7 | 1873 | 5150485 | ||

Herpud2
Genomic Map