siRNA Details
| Gene Symbol: Olfr160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030553.2 | NP_085030.2 | 1 | 626 | ||
| GenBank mRNA | BC130240.1 | - | 1 | 627 | 40142140 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr160| Gene Symbol: Olfr160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030553.2 | NP_085030.2 | 1 | 626 | ||
| GenBank mRNA | BC130240.1 | - | 1 | 627 | 40142140 | |