siRNA Details
| Gene Symbol: ACKR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020311.2 | NP_064707.1 | 2 | 1920 | ||
| XM_005246097.2 | XP_005246154.1 | 2 | 1816 | |||
| XM_005246098.3 | XP_005246155.1 | 3 | 2058 | |||
| XM_017004516.1 | XP_016860005.1 | 2 | 1811 | |||
| GenBank mRNA | AF030297.1 | - | 2 | 1758 | ||
| BC008459.1 | - | 1 | 1317 | |||
| BC036661.2 | - | 4 | 2117 | 5267661 | ||
| U67784.1 | - | 1 | 1517 | |||

ACKR3
Genomic Map