siRNA Details
| Gene Symbol: Pbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024954.1 | NP_001020125.1 | 5 | 720 | ||
| GenBank mRNA | AJ300183.1 | - | 5 | 648 | ||
| AK038786.1 | - | 324 | ||||
| AK076968.1 | - | 3 | 345 | |||
| BC032875.1 | - | 4 | 416 | 1263363 | ||

Pbx4
Genomic Map