siRNA Details
| Gene Symbol: Cwc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290740.1 | NP_001277669.1 | 8 | 1095 | ||
| NM_030560.5 | NP_085037.2 | 9 | 1153 | |||
| NM_172667.2 | NP_766255.1 | 8 | 1092 | |||
| GenBank mRNA | AK045589.1 | - | 8 | 924 | ||
| AK077961.1 | - | 9 | 981 | |||
| AK089988.1 | - | 931 | ||||
| AK129404.1 | - | 909 | ||||
| AK132001.1 | - | 8 | 1036 | |||
| AK143352.1 | - | 8 | 925 | |||
| AK149528.1 | - | 8 | 919 | |||
| AK166487.1 | - | 8 | 931 | |||
| BC003273.1 | - | 2 | 80 | |||
| BC003993.1 | - | 8 | 905 | 3492146 | ||

Cwc22
Genomic Map