siRNA Details
| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 7 | 1104 | ||
| GenBank mRNA | AK030596.1 | - | 5 | 674 | ||
| AK180627.1 | - | 3 | 239 | |||
| AK180865.1 | - | 325 | ||||
| AK191458.1 | - | 3 | 238 | |||
| AK198839.1 | - | 3 | 239 | |||
| BC004004.1 | - | 7 | 941 | 3492768 | ||
| BC058575.1 | - | 7 | 913 | 6488611 | ||

BC004004
Genomic Map