siRNA Details
| Gene Symbol: Cblc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161844.1 | NP_001155316.1 | 6, 7 | 1021 | ||
| NM_023224.5 | NP_075713.3 | 6, 7 | 1021 | |||
| GenBank mRNA | AF319956.1 | - | 6, 7 | 994 | ||
| AK010228.1 | - | 6, 7 | 1022 | |||
| BC046337.1 | - | 6, 7 | 997 | 6395806 | ||
| BC111037.1 | - | 6, 7 | 1005 | 5353413 | ||
| BE198430.1 | - | 4, 5 | 364 | |||

Cblc
Genomic Map