siRNA Details
| Gene Symbol: Ghdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031871.1 | NP_114077.1 | 7 | 1269 | ||
| GenBank mRNA | AF316996.1 | - | 7 | 1269 | ||
| AK135322.1 | - | 7 | 1271 | |||
| AK143922.1 | - | 7 | 1238 | |||
| AK161571.1 | - | 4 | 417 | |||
| BC005639.1 | - | 7 | 1250 | 3256858 | ||

Ghdc
Genomic Map