siRNA Details
| Gene Symbol: Lrba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077687.1 | NP_001071155.1 | 28 | 4773 | ||
| NM_001077688.1 | NP_001071156.1 | 28 | 4773 | |||
| NM_030695.2 | NP_109620.2 | 28 | 4773 | |||
| GenBank mRNA | CV556905.1 | - | 3 | 204 | ||

Lrba
Genomic Map