siRNA Details
| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030697.2 | NP_109622.1 | 6 | 1693 | ||
| GenBank mRNA | AK002623.1 | - | 6 | 1694 | ||
| AK132011.1 | - | 6 | 1715 | |||
| KJ650203.1 | - | 5 | 1624 | |||
| KJ663713.1 | - | 5 | 1465 | |||
| KJ663714.1 | - | 6 | 1486 | |||

Kank3
Genomic Map