siRNA Details
| Gene Symbol: Hspb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF250139.1 | - | 1, 2 | 390 | ||
| AF273453.1 | - | 1, 2 | 736 | |||
| AK005052.1 | - | 1, 2 | 758 | |||
| BC003971.1 | - | 1, 2 | 746 | |||
| BC011219.1 | - | 1, 2 | 739 | 4195285 | ||

Hspb8
Genomic Map