siRNA Details
| Gene Symbol: Fcrls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030707.3 | NP_109632.2 | 4 | 551 | ||
| GenBank mRNA | AB028174.1 | - | 4 | 582 | ||
| AF302046.1 | - | 4 | 620 | |||
| AF329486.1 | - | 4 | 552 | |||
| AK034125.1 | - | 4 | 552 | |||
| AK076123.1 | - | 4 | 585 | |||
| AY506556.1 | - | 4 | 591 | |||
| AY506557.1 | - | 4 | 591 | |||
| BC016551.1 | - | 4 | 594 | 3602803 | ||

Fcrls
Genomic Map