siRNA Details
| Gene Symbol: Erap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030711.4 | NP_109636.1 | 2 | 528 | ||
| GenBank mRNA | AB047552.1 | - | 1 | 516 | ||
| AF227511.1 | - | 1 | 504 | |||
| AK029841.1 | - | 2 | 527 | |||
| AK030329.1 | - | 2 | 529 | |||
| AK036559.1 | - | 529 | ||||
| AK081181.1 | - | 2 | 555 | |||
| AK159406.1 | - | 2 | 531 | |||
| AK163954.1 | - | 2 | 548 | |||
| BC046610.1 | - | 2 | 534 | 6397585 | ||

Erap1
Genomic Map