siRNA Details
| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030717.1 | NP_109642.1 | 5, 6 | 1225 | ||
| GenBank mRNA | AF317900.1 | - | 5, 6 | 1225 | ||
| BC046293.1 | - | 5, 6 | 1152 | 6484767 | ||
| BC147809.1 | - | 5, 6 | 1170 | 9087526 | ||

Lactb
Genomic Map