siRNA Details
| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030717.1 | NP_109642.1 | 5 | 1122 | ||
| GenBank mRNA | AF317900.1 | - | 5 | 1122 | ||
| BC046293.1 | - | 5 | 1049 | 6484767 | ||
| BC147809.1 | - | 5 | 1067 | 9087526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lactb| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030717.1 | NP_109642.1 | 5 | 1122 | ||
| GenBank mRNA | AF317900.1 | - | 5 | 1122 | ||
| BC046293.1 | - | 5 | 1049 | 6484767 | ||
| BC147809.1 | - | 5 | 1067 | 9087526 | ||