siRNA Details
| Gene Symbol: Gpr84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030720.1 | NP_109645.1 | 2 | 197 | ||
| GenBank mRNA | AF272948.1 | - | 2 | 197 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr84| Gene Symbol: Gpr84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030720.1 | NP_109645.1 | 2 | 197 | ||
| GenBank mRNA | AF272948.1 | - | 2 | 197 | ||
Set Membership: |
|