siRNA Details
| Gene Symbol: Cemip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030728.4 | NP_109653.3 | 5 | 695 | ||
| GenBank mRNA | AB103331.1 | - | 5 | 694 | ||
| AK036809.1 | - | 5 | 695 | |||
| AK162689.1 | - | 5 | 693 | |||
| BC115873.1 | - | 5 | 590 | 40086324 | ||
| BC116643.1 | - | 5 | 589 | 40086325 | ||

Cemip
Genomic Map