siRNA Details
| Gene Symbol: Rad54l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030730.2 | NP_109655.2 | 18 | 3223 | ||
| GenBank mRNA | AJ132389.1 | - | 18 | 3124 | ||
| AK143506.1 | - | 17 | 2839 | |||
| AK155151.1 | - | 18 | 3180 | |||
| BC133714.1 | - | 18 | 3072 | 40143565 | ||

Rad54l2
Genomic Map