siRNA Details
| Gene Symbol: Gpr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030733.3 | NP_109658.2 | 2 | 1348 | ||
| GenBank mRNA | AK036257.1 | - | 2 | 1349 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr63| Gene Symbol: Gpr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030733.3 | NP_109658.2 | 2 | 1348 | ||
| GenBank mRNA | AK036257.1 | - | 2 | 1349 | ||
Set Membership: |
|