siRNA Details
| Gene Symbol: Vmn1r172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030735.1 | NP_109660.1 | 2 | 1088 | ||
| GenBank mRNA | AF324873.1 | - | 2 | 1088 | ||
| BC139316.1 | - | 2 | 1083 | 8862338 | ||
| BC139317.1 | - | 2 | 1083 | 8862339 | ||
| BC145601.1 | - | 2 | 1083 | 9054167 | ||

Vmn1r172
Genomic Map