siRNA Details
Gene Symbol: Vmn1r65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030738.2 | NP_109663.1 | 3 | 1398 | ||
GenBank mRNA | AF324870.1 | - | 3 | 1399 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030738.2 | NP_109663.1 | 3 | 1398 | ||
GenBank mRNA | AF324870.1 | - | 3 | 1399 |