siRNA Details
| Gene Symbol: Ankrd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030886.2 | NP_112148.2 | 33, 34 | 7883 | ||
| NM_198010.2 | NP_932127.2 | 32, 33 | 7130 | |||
| GenBank mRNA | AK031643.1 | - | 1, 2 | 1649 | ||
| AK043407.1 | - | 4, 5 | 703 | |||
| AK076402.1 | - | 5, 6 | 2172 | |||
| AK129193.1 | - | 9, 10 | 3300 | |||
| BC039213.1 | - | 5, 6 | 2043 | |||
| BC057195.1 | - | 5, 6 | 1872 | |||

Ankrd17
Genomic Map