siRNA Details
| Gene Symbol: Gpank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128597.1 | NP_001122069.1 | 2 | 687 | ||
| NM_032460.2 | NP_115849.2 | 1 | 485 | |||
| GenBank mRNA | AB221568.1 | - | 1 | 452 | ||
| BC057902.1 | - | 2 | 646 | 5027029 | ||
| BC120637.1 | - | 1 | 486 | 40129560 | ||
| BC120639.1 | - | 1 | 486 | 40129562 | ||

Gpank1
Genomic Map