siRNA Details
| Gene Symbol: Ift122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167763.1 | NP_001161235.1 | 7 | 610 | ||
| NM_031177.4 | NP_112454.2 | 7 | 610 | |||
| GenBank mRNA | AK031308.1 | - | 7 | 498 | ||
| AK044456.1 | - | 7 | 610 | |||
| AK131115.1 | - | 7 | 537 | |||
| AK167330.1 | - | 7 | 565 | |||
| AK167412.1 | - | 7 | 565 | |||
| AK181476.1 | - | 1 | 63 | |||
| BC060621.1 | - | 7 | 697 | |||
| BC066083.1 | - | 7 | 529 | 30355922 | ||
| BC067415.1 | - | 7 | 526 | 30063124 | ||

Ift122
Genomic Map