siRNA Details
| Gene Symbol: Tmem108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178638.4 | NP_848753.1 | 4 | 432 | ||
| GenBank mRNA | AK044989.1 | - | 4 | 395 | ||
| AK047474.1 | - | 4 | 394 | |||
| AK052330.1 | - | 4 | 432 | |||
| AK077586.1 | - | 4 | 386 | |||
| AK086266.1 | - | 4 | 394 | |||
| AK132350.1 | - | 3 | 326 | |||
| AK141589.1 | - | 4 | 398 | |||
| BC052085.1 | - | 4 | 366 | 6404949 | ||

Tmem108
Genomic Map