siRNA Details
| Gene Symbol: Klb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031180.2 | NP_112457.1 | 2 | 1208 | ||
| GenBank mRNA | AF165170.1 | - | 2 | 1208 | ||
| AF178429.1 | - | 2 | 1203 | |||
| AK133823.1 | - | 2 | 1235 | |||
| BC138008.1 | - | 2 | 1297 | 8861027 | ||
| BC138010.1 | - | 2 | 1297 | 8861029 | ||

Klb
Genomic Map