siRNA Details
| Gene Symbol: Plekha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031257.3 | NP_112547.1 | 5, 6 | 518 | ||
| GenBank mRNA | AF286161.1 | - | 5, 6 | 472 | ||
| AF418551.1 | - | 4, 5 | 384 | |||
| AK032325.1 | - | 5, 6 | 528 | |||
| AK042324.1 | - | 5, 6 | 492 | |||
| AK078212.1 | - | 5, 6 | 497 | |||
| AK088395.1 | - | 5, 6 | 460 | |||
| AK153117.1 | - | 5, 6 | 490 | |||
| BC010215.1 | - | 5, 6 | 439 | 3498236 | ||

Plekha2
Genomic Map