siRNA Details
| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031375.4 | NP_113552.3 | 3 | 375 | ||
| GenBank mRNA | AB047544.1 | - | 3 | 401 | ||
| AK083330.1 | - | 2 | 768 | |||
| AK144756.1 | - | 3 | 371 | |||
| AK146059.1 | - | 2 | 750 | |||
| AK181653.1 | - | 1 | 57 | |||
| BC004034.1 | - | 362 | 3495639 | |||

Ngrn
Genomic Map